Merck
  • Regulation of S-formylglutathione hydrolase by the anti-aging gene klotho.
Regulation of S-formylglutathione hydrolase by the anti-aging gene klotho.
Oncotarget (2017-11-29)
Yuechi Xu, Zhongjie Sun
ABSTRACT
Klotho is an aging-suppressor gene. The purpose of this study is to investigate the binding sites (receptors) and function of short-form Klotho (Skl). We showed that Skl physically bound to multiple proteins. We found physical and functional interactions between Skl and
Sigma-Aldrich
MISSION® esiRNA, targeting human ZNF354C