Merck
  • Insights into the multi-scale structure and digestibility of heat-moisture treated rice starch.
Insights into the multi-scale structure and digestibility of heat-moisture treated rice starch.
Food chemistry (2017-10-19)
Hongwei Wang, Yufan Liu, Ling Chen, Xiaoxi Li, Jun Wang, Fengwei Xie
ABSTRACT
The digestibility and structural changes of rice starch induced by heat-moisture treatment (HMT) were investigated, and the relationships among the moisture content-starch structure-starch digestibility were revealed. HMT could simultaneously disorder and reassemble the rice starch molecules across multi-scale lengths and convert some fractions of rapidly-digestible starch (RDS) into slowly-digestible starch (SDS) and resistant starch (RS). In particular, the HMT rice starch with less than 30% moisture content showed a higher SDS+RS content (25.0%). During HMT, SDS and RS were preferably formed by the degraded starch molecules with M
Sigma-Aldrich
α-Amylase, heat-stable, solution, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
Amylopectin from potato starch
Sigma-Aldrich
Pancreatin from porcine pancreas, 8 × USP specifications