Merck
  • Shengjiang Xiexin Decoction Alters Pharmacokinetics of Irinotecan by Regulating Metabolic Enzymes and Transporters: A Multi-Target Therapy for Alleviating the Gastrointestinal Toxicity.
Shengjiang Xiexin Decoction Alters Pharmacokinetics of Irinotecan by Regulating Metabolic Enzymes and Transporters: A Multi-Target Therapy for Alleviating the Gastrointestinal Toxicity.
Frontiers in pharmacology (2017-11-23)
Huan-Yu Guan, Peng-Fei Li, Xiao-Ming Wang, Jia-Jing Yue, Yang He, Xiao-Mei Luo, Mei-Feng Su, Shang-Gao Liao, Yue Shi
ABSTRACT
Shengjiang Xiexin decoction (SXD), a classic traditional Chinese medical formula chronicled in
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Sigma-Aldrich
8-Cyclopentyl-1,3-dimethylxanthine, ≥98% (HPLC), powder
Sigma-Aldrich
Uridine 5′-diphosphoglucuronic acid ammonium salt, 98-100%