Merck
  • Use of a Plackett-Burman statistical design to determine the effect of selected amino acids on monoclonal antibody production in CHO cells.
Use of a Plackett-Burman statistical design to determine the effect of selected amino acids on monoclonal antibody production in CHO cells.
Biotechnol. Progress (2011)
Gonzalez-Leal IJ