Merck
  • Sox2 Communicates with Tregs Through CCL1 to Promote the Stemness Property of Breast Cancer Cells.
Sox2 Communicates with Tregs Through CCL1 to Promote the Stemness Property of Breast Cancer Cells.
Stem cells (Dayton, Ohio) (2017-10-19)
Yingxi Xu, Xiaoli Dong, Pingping Qi, Yujie Ye, Wenzhi Shen, Liang Leng, Lina Wang, Xuefei Li, Xiaohe Luo, Yanan Chen, Peiqing Sun, Rong Xiang, Na Li
ABSTRACT
As an important component of the tumor microenvironment, CD4+ CD25+ Tregs reduce antitumor immunity, promote angiogenesis and metastasis in breast cancer. However, their function in regulating the "stemness" of tumor cells and the communication between Tregs and cancer stem cells (CSCs) remain elusive. Here, we disclose that the primarily cultured Tregs isolated from breast-tumor-bearing Foxp3-EGFP mouse upregulate the stemness property of breast cancer cells. Tregs increased the side-population and the Aldehyde dehydrogenase-bright population of mouse breast cancer cells, promoted their sphere formation in a paracrine manner, and enhanced the expression of stemness genes, such as Sox2 and so forth. In addition, Tregs increased tumorigenesis, metastasis, and chemoresistance of breast cancer cells. Furthermore, Sox2-overexpression tumor cells activated NF-κB-CCL1 signaling to recruit Tregs through reducing the binding of H3K27Me3 on promoter regions of p65 and Ccl1. These findings reveal the functional interaction between Tregs and CSCs and indicate that targeting on the communication between them is a promising strategy in breast cancer therapy. Stem Cells 2017;35:2351-2365.