Merck
  • ASXL3 Is a Novel Pluripotency Factor in Human Respiratory Epithelial Cells and a Potential Therapeutic Target in Small Cell Lung Cancer.
ASXL3 Is a Novel Pluripotency Factor in Human Respiratory Epithelial Cells and a Potential Therapeutic Target in Small Cell Lung Cancer.
Cancer research (2017-09-25)
Vivek Shukla, Mahadev Rao, Hongen Zhang, Jeanette Beers, Darawalee Wangsa, Danny Wangsa, Floryne O Buishand, Yonghong Wang, Zhiya Yu, Holly S Stevenson, Emily S Reardon, Kaitlin C McLoughlin, Andrew S Kaufman, Eden C Payabyab, Julie A Hong, Mary Zhang, Sean Davis, Daniel Edelman, Guokai Chen, Markku M Miettinen, Nicholas P Restifo, Thomas Ried, Paul A Meltzer, David S Schrump
ABSTRACT
In this study, we generated induced pluripotent stem cells (iPSC) from normal human small airway epithelial cells (SAEC) to investigate epigenetic mechanisms of stemness and pluripotency in lung cancers. We documented key hallmarks of reprogramming in lung iPSCs (Lu-iPSC) that coincided with modulation of more than 15,000 genes relative to parental SAECs. Of particular novelty, we identified the PRC2-associated protein, ASXL3, which was markedly upregulated in Lu-iPSCs and small cell lung cancer (SCLC) lines and clinical specimens. ASXL3 overexpression correlated with increased genomic copy number in SCLC lines. ASXL3 silencing inhibited proliferation, clonogenicity, and teratoma formation by Lu-iPSCs, and diminished clonogenicity and malignant growth of SCLC cells in vivo Collectively, our studies validate the utility of the Lu-iPSC model for elucidating epigenetic mechanisms contributing to pulmonary carcinogenesis and highlight ASXL3 as a novel candidate target for SCLC therapy. Cancer Res; 77(22); 6267-81. ©2017 AACR.
Sigma-Aldrich
Anti-Actin, α-Smooth Muscle antibody, Mouse monoclonal, clone 1A4, purified from hybridoma cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium bisulfite solution, purum, ~40%
Pricing and availability is not currently available.
Sigma-Aldrich
Mouse IgG2b Negative Control, clone GC198, FITC conjugate, Mouse IgG2b Negative Control Monoclonal Antibody validated for use in Flow Cytometry & Immunofluorescence.
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Oct-4 Antibody, clone 10H11.2, Alexa Fluor 488 conjugate, clone 10H11.2, from mouse, ALEXA FLUOR 488
Pricing and availability is not currently available.
Sigma-Aldrich
Milli-Mark® Anti-Nanog-Alexa Fluor 488 Antibody, NT, Milli-Mark®, from rabbit
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-TRA-1-60 Antibody, clone TRA-1-60, FITC conjugate, clone TRA-1-60, from mouse, FITC conjugate
Pricing and availability is not currently available.