Merck
A novel vector for transgenesis in the rat CNS.
Acta neuropathologica communications (2017-11-22)
T Peter Lopez, Kurt Giles, Brittany N Dugger, Abby Oehler, Carlo Condello, Zuzana Krejciova, Julian A Castaneda, George A Carlson, Stanley B Prusiner
ABSTRACT
The larger brain of the rat enables a much greater repertoire of complex behaviors than mice, likely making rats preferential for investigating neurodegeneration. Because molecular tools for specific expression of transgenes in the rat brain are sparse, we chose Prnp encoding the prion protein (PrP) to develop a novel vector to drive transgene expression in the rat brain. We compared the rat Prnp sequence with mouse and Syrian hamster Prnp sequences, identifying conserved genetic elements and hypothesizing that these elements would be able to drive neuronal transgene expression. We investigated this by generating a vector termed RaPrnp that encompasses portions of the rat Prnp gene. Importantly, we replaced the rat Prnp open reading frame (ORF) with a cloning site for rapid and seamless In-Fusion cloning. To validate the in vivo neuronal specificity of the RaPrnp vector in rats, we generated stable RaPrnp-LacZ/enhanced green fluorescent protein (EGFP) transgenic (Tg) rat lines, which led to robust LacZ activity and high EGFP fluorescence in the central nervous system of embryos and adult animals. Next, we restored the rat Prnp ORF and generated multiple Tg(RaPrnp-PrP) lines, demonstrating that overexpression of Prnp accelerates the onset of scrapie. While the incubation time in wild-type (WT) rats was 175 ± 3 days post inoculation (dpi), one line, Tg2919, overexpressed RaPrP
Sigma-Aldrich
M16 Medium, With sodium bicarbonate and lactic acid, without penicillin and streptomycin, liquid, sterile-filtered
Sigma-Aldrich
M2 medium, With HEPES, without penicillin and streptomycin, liquid, sterile-filtered, suitable for mouse embryo cell culture
Sigma-Aldrich
Hyaluronidase from bovine testes, Type IV-S, powder, suitable for mouse embryo cell culture, 750-3000 units/mg solid