Merck
Moderate O
Free radical research (2017-10-13)
Parvaneh Nasezadeh, Farshad Shahi, Mohammadjavad Fridoni, Enayatollah Seydi, Morteza Izadi, Ahmad Salimi
ABSTRACT
Mitochondrial damage and oxidative stress are known to contribute to the pathogenesis of noise-induced hearing loss (NIHL). In this study, we examined the protective effect of O
Sigma-Aldrich
ADP/ATP Ratio Assay Kit, sufficient for 100 tests (bioluminescent)
Pricing and availability is not currently available.
Sigma-Aldrich
Pentostatin, ≥95% (HPLC)
Pricing and availability is not currently available.