Merck
  • Glut3 Addiction Is a Druggable Vulnerability for a Molecularly Defined Subpopulation of Glioblastoma.
Glut3 Addiction Is a Druggable Vulnerability for a Molecularly Defined Subpopulation of Glioblastoma.
Cancer cell (2017-12-05)
Érika Cosset, Sten Ilmjärv, Valérie Dutoit, Kathryn Elliott, Tami von Schalscha, Maria F Camargo, Alexander Reiss, Toshiro Moroishi, Laetitia Seguin, German Gomez, Jung-Soon Moo, Olivier Preynat-Seauve, Karl-Heinz Krause, Hervé Chneiweiss, Jann N Sarkaria, Kun-Liang Guan, Pierre-Yves Dietrich, Sara M Weis, Paul S Mischel, David A Cheresh
ABSTRACT
While molecular subtypes of glioblastoma (GBM) are defined using gene expression and mutation profiles, we identify a unique subpopulation based on addiction to the high-affinity glucose transporter, Glut3. Although Glut3 is a known driver of a cancer stem cell phenotype, direct targeting is complicated by its expression in neurons. Using established GBM lines and patient-derived stem cells, we identify a subset of tumors within the "proneural" and "classical" subtypes that are addicted to aberrant signaling from integrin αvβ3, which activates a PAK4-YAP/TAZ signaling axis to enhance Glut3 expression. This defined subpopulation of GBM is highly sensitive to agents that disrupt this pathway, including the integrin antagonist cilengitide, providing a targeted therapeutic strategy for this unique subset of GBM tumors.
Sigma-Aldrich
MISSION® esiRNA, targeting human SLC2A3
Sigma-Aldrich
Verteporfin, ≥94% (HPLC)
Sigma-Aldrich
3-(Biphenyl-4-yl)-5-(4-tert-butylphenyl)-4-phenyl-4H-1,2,4-triazole, 97%
Sigma-Aldrich
MISSION® esiRNA, targeting human PAK4