Merck
  • RNA polymerase II primes Polycomb-repressed developmental genes throughout terminal neuronal differentiation.
RNA polymerase II primes Polycomb-repressed developmental genes throughout terminal neuronal differentiation.
Molecular systems biology (2017-10-19)
Carmelo Ferrai, Elena Torlai Triglia, Jessica R Risner-Janiczek, Tiago Rito, Owen Jl Rackham, Inês de Santiago, Alexander Kukalev, Mario Nicodemi, Altuna Akalin, Meng Li, Mark A Ungless, Ana Pombo
ABSTRACT
Polycomb repression in mouse embryonic stem cells (ESCs) is tightly associated with promoter co-occupancy of RNA polymerase II (RNAPII) which is thought to prime genes for activation during early development. However, it is unknown whether RNAPII poising is a general feature of Polycomb repression, or is lost during differentiation. Here, we map the genome-wide occupancy of RNAPII and Polycomb from pluripotent ESCs to non-dividing functional dopaminergic neurons. We find that poised RNAPII complexes are ubiquitously present at Polycomb-repressed genes at all stages of neuronal differentiation. We observe both loss and acquisition of RNAPII and Polycomb at specific groups of genes reflecting their silencing or activation. Strikingly, RNAPII remains poised at transcription factor genes which are silenced in neurons through Polycomb repression, and have major roles in specifying other, non-neuronal lineages. We conclude that RNAPII poising is intrinsically associated with Polycomb repression throughout differentiation. Our work suggests that the tight interplay between RNAPII poising and Polycomb repression not only instructs promoter state transitions, but also may enable promoter plasticity in differentiated cells.
Sigma-Aldrich
Picrotoxin, powder
Sigma-Aldrich
NBQX hydrate, powder, ≥98% (HPLC)
Sigma-Aldrich
Anti-GABA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Vesicular Glutamate Transporter 2 Antibody, clone 8G9.2, Chemicon®, from mouse
Sigma-Aldrich
Monoclonal Anti-β-Tubulin III antibody produced in mouse, clone SDL.3D10, ascites fluid
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Standardized vial containing 2,000 Kunitz units of DNase I (D4527), vial of ≥0.25 mg total protein
Sigma-Aldrich
UNC1999, ≥98% (HPLC)
Sigma-Aldrich
Anti-trimethyl-Histone H3 (Lys27) Antibody, Upstate®, from rabbit