Merck
  • Isolation, characterization, and differentiation of multipotent neural
    progenitor cells from human cerebrospinal fluid in fetal
    cystic myelomeningocele
Isolation, characterization, and differentiation of multipotent neural<BR/> progenitor cells from human cerebrospinal fluid in fetal<BR/> cystic myelomeningocele
Stem Cell Research (2017)
Mario Marotta