Merck
  • Seed-mediated growth of Au@Ag core-shell nanorods for the detection of ellagic acid in whitening cosmetics.
Seed-mediated growth of Au@Ag core-shell nanorods for the detection of ellagic acid in whitening cosmetics.
Analytica chimica acta (2018-01-08)
Yi Wang, Yang Zeng, Wensheng Fu, Pu Zhang, Ling Li, Cuiying Ye, Lan Yu, Xiaochun Zhu, Song Zhao
ABSTRACT
Seed-mediated growth has been employed as a simple and powerful means to the shape-controlled synthesis of metal nanocrystals. In this work, we apply the principle of seed-mediated growth in analytical chemistry, and achieve improved sensitivity due to the low energy barrier in the target-induced formation of bimetallic nanoparticles with core-shell structure. As a result, a simple, reliable, highly sensitive and selective method for the detection of ellagic acid (EA), a naturally occurring polyphenolic antioxidant, has been developed. With the aid of EA in alkaline solution, Ag
Sigma-Aldrich
5-Bromosalicylic acid, technical grade, 90%
Pricing and availability is not currently available.