Merck
  • The correlation between concentrations of zolpidem and benzodiazepines in segmental hair samples and use patterns.
The correlation between concentrations of zolpidem and benzodiazepines in segmental hair samples and use patterns.
Forensic science international (2017-11-15)
Hyojeong Kim, Sangeun Lee, Sanghwan In, Meejung Park, Sungnam Cho, Junguk Shin, Hunjoo Lee, Eunyoung Han
ABSTRACT
The aim of this study was to investigate the correlation between histories of zolpidem and benzodiazepines use and their concentrations in hair as determined by segmental hair analysis, that is, by analyzing hair samples taken 0-1, 1-2, 2-3, 3-4, 4-5, and 5-6cm etc. and 0-3cm from the scalp, and whole hair. Of the 23 hair samples examined, 18 were collected from patients in a rehabilitation program and five were from patients that had taken zolpidem only once by prescription. All 23 patients provided written informed consent after reviewing the research plan, described their zolpidem and benzodiazepines use histories accurately, and provided hair samples, which were weighed, washed, cut into lengths of <1mm, and extracted in 100% methanol for 16h (diazepam-d