Merck
  • WITHDRAWN: Regulation of Pyruvate Kinase M Switch towards PKM1 by LKB1-AMPK Axis Tolerates Hypoglycemic Stress in Cancer Cells.
WITHDRAWN: Regulation of Pyruvate Kinase M Switch towards PKM1 by LKB1-AMPK Axis Tolerates Hypoglycemic Stress in Cancer Cells.
The Journal of biological chemistry (2016-04-30)
Prakasam Gopinath, Rajnish Kumar Singh, Mohammad Askandar Iqbal, Ashu Bhan Tiku, Rameshwar N K Bamezai
ABSTRACT
This article has been withdrawn by the authors. The PKM2 immunoblot in Fig 2E was reused as part of the Caspase-3 immunoblot in Fig 9C. The PKM2 immunoblot from 5 mM Glu, fractions 1-10 was reused as the PKM2 immunoblot from 1 mM Glu, fractions 1-10. The actin immunoblot from A549 cells from Fig 5A was reused as the actin blot from Fig 7C.
Sigma-Aldrich
Anti-PKM2 (isoform M1) antibody produced in rabbit, ~1.5 mg/mL, affinity isolated antibody
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture