Merck
  • Modulating Protein-Protein Interactions of the Mitotic Polo-like Kinases to Target Mutant KRAS.
Modulating Protein-Protein Interactions of the Mitotic Polo-like Kinases to Target Mutant KRAS.
Cell chemical biology (2017-08-16)
Ana J Narvaez, Suzan Ber, Alex Crooks, Amy Emery, Bryn Hardwick, Estrella Guarino Almeida, David J Huggins, David Perera, Meredith Roberts-Thomson, Roberta Azzarelli, Fiona E Hood, Ian A Prior, David W Walker, Richard Boyce, Robert G Boyle, Samuel P Barker, Christopher J Torrance, Grahame J McKenzie, Ashok R Venkitaraman
ABSTRACT
Mutations activating KRAS underlie many forms of cancer, but are refractory to therapeutic targeting. Here, we develop Poloppin, an inhibitor of protein-protein interactions via the Polo-box domain (PBD) of the mitotic Polo-like kinases (PLKs), in monotherapeutic and combination strategies to target mutant KRAS. Poloppin engages its targets in biochemical and cellular assays, triggering mitotic arrest with defective chromosome congression. Poloppin kills cells expressing mutant KRAS, selectively enhancing death in mitosis. PLK1 or PLK4 depletion recapitulates these cellular effects, as does PBD overexpression, corroborating Poloppin's mechanism of action. An optimized analog with favorable pharmacokinetics, Poloppin-II, is effective against KRAS-expressing cancer xenografts. Poloppin resistance develops less readily than to an ATP-competitive PLK1 inhibitor; moreover, cross-sensitivity persists. Poloppin sensitizes mutant KRAS-expressing cells to clinical inhibitors of c-MET, opening opportunities for combination therapy. Our findings exemplify the utility of small molecules modulating the protein-protein interactions of PLKs to therapeutically target mutant KRAS-expressing cancers.
Sigma-Aldrich
Nocodazole, ≥99% (TLC), powder
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-74, ascites fluid
Sigma-Aldrich
Poloppin, ≥98% (HPLC)