Merck
  • Alzheimer's disease-causing proline substitutions lead to presenilin 1 aggregation and malfunction.
Alzheimer's disease-causing proline substitutions lead to presenilin 1 aggregation and malfunction.
The EMBO journal (2015-10-07)
Tziona Ben-Gedalya, Lorna Moll, Michal Bejerano-Sagie, Samuel Frere, Wayne A Cabral, Dinorah Friedmann-Morvinski, Inna Slutsky, Tal Burstyn-Cohen, Joan C Marini, Ehud Cohen
ABSTRACT
Do different neurodegenerative maladies emanate from the failure of a mutual protein folding mechanism? We have addressed this question by comparing mutational patterns that are linked to the manifestation of distinct neurodegenerative disorders and identified similar neurodegeneration-linked proline substitutions in the prion protein and in presenilin 1 that underlie the development of a prion disorder and of familial Alzheimer's disease (fAD), respectively. These substitutions were found to prevent the endoplasmic reticulum (ER)-resident chaperone, cyclophilin B, from assisting presenilin 1 to fold properly, leading to its aggregation, deposition in the ER, reduction of γ-secretase activity, and impaired mitochondrial distribution and function. Similarly, reduced quantities of the processed, active presenilin 1 were observed in brains of cyclophilin B knockout mice. These discoveries imply that reduced cyclophilin activity contributes to the development of distinct neurodegenerative disorders, propose a novel mechanism for the development of certain fAD cases, and support the emerging theme that this disorder can stem from aberrant presenilin 1 function. This study also points at ER chaperones as targets for the development of counter-neurodegeneration therapies.
Sigma-Aldrich
Monoclonal Anti-Vimentin antibody produced in mouse, clone V9, ascites fluid
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Millipore
High Sensitivity Human Amyloid β40 ELISA
Millipore
High Sensitivity Human Amyloid β42 ELISA
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, BioReagent, Molecular Biology, ≥95%
Millipore
Protease Inhibitor Cocktail Set III, EDTA-Free, Protease inhibitor cocktail III, EDTA-free for inhibiting aspartic, cysteine, and serine proteases as well as aminopeptidases in mammalian cells and tissues.
Sigma-Aldrich
Anti-Presenilin-1 Antibody, NT, clone hPS1-NT, culture supernatant, clone hPS1-NT, Chemicon®