Merck
  • Disruptions in asymmetric centrosome inheritance and WDR62-Aurora kinase B interactions in primary microcephaly.
Disruptions in asymmetric centrosome inheritance and WDR62-Aurora kinase B interactions in primary microcephaly.
Scientific reports (2017-03-09)
Paraskevi Sgourdou, Ketu Mishra-Gorur, Ichiko Saotome, Octavian Henagariu, Beyhan Tuysuz, Cynthia Campos, Keiko Ishigame, Krinio Giannikou, Jennifer L Quon, Nenad Sestan, Ahmet O Caglayan, Murat Gunel, Angeliki Louvi
ABSTRACT
Recessive mutations in WD repeat domain 62 (WDR62) cause microcephaly and a wide spectrum of severe brain malformations. Disruption of the mouse ortholog results in microcephaly underlain by reduced proliferation of neocortical progenitors during late neurogenesis, abnormalities in asymmetric centrosome inheritance leading to neuronal migration delays, and altered neuronal differentiation. Spindle pole localization of WDR62 and mitotic progression are defective in patient-derived fibroblasts, which, similar to mouse neocortical progenitors, transiently arrest at prometaphase. Expression of WDR62 is closely correlated with components of the chromosome passenger complex (CPC), a key regulator of mitosis. Wild type WDR62, but not disease-associated mutant forms, interacts with the CPC core enzyme Aurora kinase B and staining of CPC components at centromeres is altered in patient-derived fibroblasts. Our findings demonstrate critical and diverse functions of WDR62 in neocortical development and provide insight into the mechanisms by which its disruption leads to a plethora of structural abnormalities.
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
ApopTag Peroxidase In Situ Apoptosis Detection Kit, The ApopTag Peroxidase In Situ Apoptosis Detection Kit detects apoptotic cells in situ by labeling & detecting DNA strand breaks by the TUNEL method.
Sigma-Aldrich
Anti-V5 Epitope Tag Antibody, Chemicon®, from rabbit