Merck
  • A novel oncogenic BTK isoform is overexpressed in colon cancers and required for RAS-mediated transformation.
A novel oncogenic BTK isoform is overexpressed in colon cancers and required for RAS-mediated transformation.
Oncogene (2016-01-26)
E Grassilli, F Pisano, A Cialdella, S Bonomo, C Missaglia, M G Cerrito, L Masiero, L Ianzano, F Giordano, V Cicirelli, R Narloch, F D'Amato, B Noli, G L Ferri, B E Leone, G Stanta, S Bonin, K Helin, R Giovannoni, M Lavitrano
ABSTRACT
Bruton's tyrosine kinase (BTK) is essential for B-cell proliferation/differentiation and it is generally believed that its expression and function are limited to bone marrow-derived cells. Here, we report the identification and characterization of p65BTK, a novel isoform abundantly expressed in colon carcinoma cell lines and tumour tissue samples. p65BTK protein is expressed, through heterogeneous nuclear ribonucleoprotein K (hnRNPK)-dependent and internal ribosome entry site-driven translation, from a transcript containing an alternative first exon in the 5'-untranslated region, and is post-transcriptionally regulated, via hnRNPK, by the mitogen-activated protein kinase (MAPK) pathway. p65BTK is endowed with strong transforming activity that depends on active signal-regulated protein kinases-1/2 (ERK1/2) and its inhibition abolishes RAS transforming activity. Accordingly, p65BTK overexpression in colon cancer tissues correlates with ERK1/2 activation. Moreover, p65BTK inhibition affects growth and survival of colon cancer cells. Our data reveal that BTK, via p65BTK expression, is a novel and powerful oncogene acting downstream of the RAS/MAPK pathway and suggest that its targeting may be a promising therapeutic approach.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Magna RIP® RNA-Binding Protein Immunoprecipitation Kit, RNA Immunoprecipitation (RIP) Kit containing all necessary reagents to perform 12 individual RNA-binding protein immunoprecipitation (RIP) reactions using protein A/G magnetic beads.
Sigma-Aldrich
Anti-phospho-hnRNP K (pSer284) antibody produced in rabbit, affinity isolated antibody
Sigma-Aldrich
Anti-Vinculin antibody, Mouse monoclonal, clone hVIN-1, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Ras Antibody, clone RAS10, clone RAS10, Upstate®, from mouse