Merck
  • Identification by shape-based virtual screening and evaluation of new tyrosinase inhibitors.
Identification by shape-based virtual screening and evaluation of new tyrosinase inhibitors.
PeerJ (2018-02-01)
Qi Li, Hongyu Yang, Jun Mo, Yao Chen, Yue Wu, Chen Kang, Yuan Sun, Haopeng Sun
ABSTRACT
Targeting tyrosinase is considered to be an effective way to control the production of melanin. Tyrosinase inhibitor is anticipated to provide new therapy to prevent skin pigmentation, melanoma and neurodegenerative diseases. Herein, we report our results in identifying new tyrosinase inhibitors. The shape-based virtual screening was performed to discover new tyrosinase inhibitors. Thirteen potential hits derived from virtual screening were tested by biological determinations. Compound 5186-0429 exhibited the most potent inhibitory activity. It dose-dependently inhibited the activity of tyrosinase, with the IC
Sigma-Aldrich
Tyrosinase from mushroom, lyophilized powder, ≥1000 unit/mg solid