Merck
  • Core Richness of N-Glycans of Caenorhabditis elegans: A Case Study on Chemical and Enzymatic Release.
Core Richness of N-Glycans of Caenorhabditis elegans: A Case Study on Chemical and Enzymatic Release.
Analytical chemistry (2017-11-29)
Shi Yan, Jorick Vanbeselaere, Chunsheng Jin, Markus Blaukopf, Florian Wöls, Iain B H Wilson, Katharina Paschinger
ABSTRACT
Despite years of research, the glycome of the model nematode Caenorhabditis elegans is still not fully understood. Certainly, data over the years have indicated that this organism synthesizes unusual N-glycans with a range of galactose and fucose modifications on the Man