Merck
Optimization of
Oncotarget (2017-12-22)
Marius Ilie, Catherine Butori, Sandra Lassalle, Simon Heeke, Nicolas Piton, Jean-Christophe Sabourin, Virginie Tanga, Kevin Washetine, Elodie Long-Mira, Priscilla Maitre, Nathalie Yazbeck, Olivier Bordone, Virginie Lespinet, Sylvie Leroy, Charlotte Cohen, Jérôme Mouroux, Charles Hugo Marquette, Véronique Hofman, Paul Hofman
ABSTRACT
Treatment with EGFR inhibitors is limited to patients with advanced/metastatic non-small cell lung cancer who have known