- L-cysteine functionalized gold nanoparticles for the colorimetric detection of Hg2+ induced by ultraviolet light
L-cysteine functionalized gold nanoparticles for the colorimetric detection of Hg2+ induced by ultraviolet light
Nanotechnology (2009)
Chai F