Merck
  • Structure of a triclinic ternary complex of horse liver alcohol dehydrogenase at 2.9 A resolution.
Structure of a triclinic ternary complex of horse liver alcohol dehydrogenase at 2.9 A resolution.
Journal of molecular biology (1981-03-15)
H Eklund, J P Samma, L Wallén, C I Brändén, A Akeson, T A Jones
Sigma-Aldrich
Alcohol Dehydrogenase, recombinant from E. coli, ≥500 U/mL
Sigma-Aldrich
Alcohol Dehydrogenase equine, recombinant, expressed in E. coli, ≥0.5 U/mg