Merck
  • Cell density dependence of cAMP and cGMP levels in four human cell lines derived from carcinomas of the uterine cervix.
Cell density dependence of cAMP and cGMP levels in four human cell lines derived from carcinomas of the uterine cervix.
Gynecologic oncology (1994-03-01)
A Orbo, R Jaeger, G Sager
ABSTRACT
Several studies have shown altered biokinetics of cyclic nucleotides in human cancer. In order to mimic the growing tumor bulk in carcinomas of the uterine cervix, four human cell lines (C4-I, C33A, ME-180, and SiHa) were expanded in serum-supplemented cell cultures. The extra- and intracellular levels of cAMP and cGMP were determined at increasing cell densities. In all of the cell lines, a cell density-dependent increase in the extracellular cGMP/cAMP ratio was observed. cAMP was distributed to the extracellular compartment against a concentration gradient at low cell densities but was retained in the intracellular compartment at high cell densities. In contrast, cGMP was distributed to the extracellular compartment against a concentration gradient for the whole range of cell densities. This study suggests that the cell density-dependent increase in the extracellular cGMP/cAMP ratio in cell lines derived from carcinomas of the uterine cervix is a result of changes occurring in both intracellular levels and cellular excretion of cyclic nucleotides.
Product Description
Supelco
ORBO 80 Isocyanate Filter, with cassettes: no, filter O.D. 37 mm, pkg of 25 ea
Supelco
ORBO 52 Small Activated Silica Gel (20/40) 150/75 mg, pk 50, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 43 Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 100/50 mg, W,W,W (specially treated) separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 92 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group Vinyl acetate
Supelco
ORBO 90 Carboxen® 564 (20/45), 160/80 mg, W,F,F separators, O.D. × L 6 mm × 75 mm, pkg of 25 ea, for analyte group MEK (methylethyl ketone)
Supelco
ORBO 65P (OVS) Amberlite XAD®-4 specially cleaned, 160/80 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS), pkg of 10 ea
Supelco
ORBO 24 2-HMP on Amberlite XAD®-2 (20/40), 150/75 mg, W,W,W separators, O.D. × L 6 mm × 105 mm, pkg of 25 ea
Supelco
ORBO 609 Amberlite XAD®-2 (20/50) 400/200 mg, W,W,W separators, O.D. × L 8 mm × 110 mm, pkg of 50 ea
Supelco
ORBO 402 Tenax® TA specially treated (35/60), 100/50 mg, W,W,W separators, O.D. × L 8 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 554 H2SO4 on Silica Gel (20/40) 150/75 mg, W,W,W separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea
Supelco
ORBO 53 Activated Silica Gel (20/40) specially cleaned, 400/200 mg with Glass Fiber Filters, GFF,F,F separators, O.D. × L 7 mm × 100 mm, pkg of 50 ea
Supelco
ORBO 23 2-HMP on Amberlite XAD®-2 (20/40), 120/60 mg, W,W,W separators, O.D. × L 6 mm × 85 mm, pkg of 25 ea
Supelco
ORBO 80 Coated Filter, with cassettes unassembled
Supelco
ORBO 49P (OVS) Supelpak-20 specially treated Amberlite XAD®-2 (20/40), 270/140 mg, RC,GFF,F,F (specially cleaned) separators, O.D. × L 13-8 mm × 75 mm , OSHA Versatile Sampler (OVS) Tube, pkg of 10 ea
Supelco
ORBO 502 Activated Silica Gel (20/45) 100/50 mg, W,W,F separators, O.D. × L 6 mm × 75 mm, pkg of 50 ea