Merck
  • Systematic structure modifications of imidazo[1,2-a]pyrimidine to reduce metabolism mediated by aldehyde oxidase (AO).
Systematic structure modifications of imidazo[1,2-a]pyrimidine to reduce metabolism mediated by aldehyde oxidase (AO).
Journal of medicinal chemistry (2011-10-01)
Angelica Linton, Ping Kang, Martha Ornelas, Susan Kephart, Qiyue Hu, Mason Pairish, Ying Jiang, Chuangxing Guo
ABSTRACT
N-{trans-3-[(5-Cyano-6-methylpyridin-2-yl)oxy]-2,2,4,4-tetramethylcyclobutyl}imidazo[1,2-a]pyrimidine-3-carboxamide (1) was recently identified as a full antagonist of the androgen receptor, demonstrating excellent in vivo tumor growth inhibition in castration-resistant prostate cancer (CRPC). However, the imidazo[1,2-a]pyrimidine moiety is rapidly metabolized by aldehyde oxidase (AO). The present paper describes a number of medicinal chemistry strategies taken to avoid the AO-mediated oxidation of this particular system. Guided by an AO protein structure-based model, our investigation revealed the most probable site of AO oxidation and the observation that altering the heterocycle or blocking the reactive site are two of the more effective strategies for reducing AO metabolism. These strategies may be useful for other drug discovery programs.
Product Description
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Supelco
Discovery® Cyano Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Supelco
Discovery® Cyano HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm