Merck
HomeGene Expression & SilencingCustom siRNA Libraries & Panels
Custom siRNA Libraries & Panels
MISSION® Custom siRNA Libraries & Panels are the most compelling collections for high-throughput screening of therapeutic-value target genes. The flexible format facilitates research for scientists who are interested in specific classes of genes as well as those who need to generate information across the entire druggable genome.
Product Benefits
  • Duplexes designed using the Rosetta Algorithm, one of the most powerful tools for high target specificity and efficient gene knockdown
  • Up to 3 duplexes available per gene, thereby allowing for optional pooling
  • Amounts of material provided allow for multiple screenings and hit follow-up
  • Gene targets picked in collaboration with major pharmaceutical company
Product Features
  • 21mer siRNA duplexes with dTdT overhangs
  • Duplex amounts provided vary (see list below)
  • 1 to 3 duplexes available per gene
  • Duplexes formatted in multiple 96-well plates
    o   80 duplexes per plate
    o   First/last columns empty
    o   Dry
  • Items provided electronically, include:
    o   Plate maps
    o   Target gene list
    o   Sequence information
  • Controls may be available (inquire)
Available Libraries & Panels
Select the library or panel of interest and then use the button above to complete the quote request form. To learn if controls are included as well as the list of genes in the library or panel of interest, please send an email to [email protected].
Library /
Panel No.
Product NameNumber of TargetsDuplex Amount
Human
1MISSION® siRNA Human Druggable Genome Library66551 nmol
2MISSION® Human Gene Family Panels Ligase Panel9491 nmol
3MISSION® siRNA Human Gene Family Panels Human Growth Factors/Receptors Panel3751 nmol
4MISSION® siRNA Human Gene Family Panels Cell Adhesion and Cytoskeleton Panel4961 nmol
5MISSION® siRNA Human Gene Family Panels Ion Channel and Transporters Panel6391 nmol
6MISSION® siRNA Human Gene Family Panels Assorted Function Panel2281 nmol
7MISSION® siRNA Human Gene Family Panels GPCR Panel3041 nmol
8MISSION® siRNA Human Gene Family Panels Hydrolase Panel2041 nmol
9MISSION® siRNA Human Gene Family Panels Metabolism and Cell Traffic Panel2171 nmol
10MISSION® Human Nucleic Acid Binding Panel3101 nmol
11MISSION® siRNA Human Gene Family Panels Oxidoreductase Panel3381 nmol
12Human Gene Family Panels Protease Panel3791 nmol
13MISSION® siRNA Human Gene Family Panels Cell Regulation Panel2271 nmol
14MISSION® siRNA Human Gene Family Panels Transfer and Carrier Proteins Panel711 nmol
15MISSION® siRNA Human Gene Family Panels Transferase Panel1841 nmol
16MISSION® siRNA Human Gene Family Set Kinase Panel7190.25 nmol
17MISSION® siRNA Human Phosphatase Panel3030.25 nmol
Mouse
18MISSION® siRNA Mouse Kinase Panel6230.5 nmol
Rat   
19MISSION® siRNA Rat Druggable Genome Library60161 nmol
20MISSION® siRNA Rat Gene Family Panels Ligase Panel8161 nmol
21MISSION® siRNA Rat Gene Family Panels Kinase Panel6691 nmol
22MISSION® siRNA Rat Gene Family Panels Phosphatase Panel2661 nmol
23MISSION® siRNA Rat Gene Family Panels Growth Factors/Receptors Panel3421 nmol
24MISSION® siRNA Rat Gene Family Panels Cell Adhesion and Cytoskeleton Panel4331 nmol
25MISSION® siRNA Rat Gene Family Panels Ion Channel and Transporters Panel5941 nmol
26MISSION® siRNA Rat Gene Family Panels Assorted Function Panel2101 nmol
27MISSION® siRNA Rat Gene Family Panels GPCR Panel2751 nmol
28MISSION® siRNA Rat Gene Family Panels Hydrolase Panel1881 nmol
29MISSION® siRNA Rat Gene Family Panels Metabolism and Cell Traffic Panel2011 nmol
30MISSION® siRNA Rat Gene Family Panels Nucleic Acid Binding Panel2871 nmol
31MISSION® siRNA Rat Gene Family Panels Oxidoreductase Panel3021 nmol
32MISSION® siRNA Rat Gene Family Panels Protease Panel3411 nmol
33MISSION® siRNA Rat Gene Family Panels Cell Surface and Nuclear Receptors Panel6471 nmol
34MISSION® siRNA Rat Gene Family Panels Cell Regulation Panel2141 nmol
35MISSION® siRNA Rat Gene Family Panels Transfer and Carrier Proteins Panel661 nmol
36MISSION® siRNA Rat Gene Family Panels Transferase Panel1741 nmol
References
1.
Hoorn EJ, Meima ME. 2012. Predicting kinase-substrate interactions in the era of proteomics. Focus on "Identifying protein kinase target preferences using mass spectrometry". American Journal of Physiology-Cell Physiology. 303(7):C711-C712. https://doi.org/10.1152/ajpcell.00232.2012
2.
Meng W, Mushika Y, Ichii T, Takeichi M. 2008. Anchorage of Microtubule Minus Ends to Adherens Junctions Regulates Epithelial Cell-Cell Contacts. Cell. 135(5):948-959. https://doi.org/10.1016/j.cell.2008.09.040
3.
Matsubara T, Kida K, Yamaguchi A, Hata K, Ichida F, Meguro H, Aburatani H, Nishimura R, Yoneda T. 2008. BMP2 Regulates Osterix through Msx2 and Runx2 during Osteoblast Differentiation. Journal of Biological Chemistry. 283(43):29119-29125. https://doi.org/10.1074/jbc.m801774200
4.
2015. Host Factors in Retroviral Integration and the Selection of Integration Target Sites.1035-1050. https://doi.org/10.1128/microbiolspec.mdna3-0026-2014
5.
Zhang XD. 2008. Genome-wide screens for effective siRNAs through assessing the size of siRNA effects. BMC Research Notes. 1(1):33. https://doi.org/10.1186/1756-0500-1-33
6.
Linsley PS, Jackson AL. 2010. Hidden reach of the micromanagers. BMC Biol. 8(1): https://doi.org/10.1186/1741-7007-8-53
7.
Majercak J, Ray WJ, Espeseth A, Simon A, Shi X, Wolffe C, Getty K, Marine S, Stec E, Ferrer M, et al. 2006. LRRTM3 promotes processing of amyloid-precursor protein by BACE1 and is a positional candidate gene for late-onset Alzheimer's disease. Proceedings of the National Academy of Sciences. 103(47):17967-17972. https://doi.org/10.1073/pnas.0605461103