Merck
  • A signal-amplifiable biochip quantifies extracellular vesicle-associated RNAs for early cancer detection.
A signal-amplifiable biochip quantifies extracellular vesicle-associated RNAs for early cancer detection.
Nature communications (2017-11-23)
Jiaming Hu, Yan Sheng, Kwang Joo Kwak, Junfeng Shi, Bohao Yu, L James Lee
ZUSAMMENFASSUNG
Detection of extracellular vesicle (EV)-associated RNAs with low expression levels in early-stage cancer remains a challenge and is highly valuable. Here, we report a nanoparticle-based biochip that could capture circulating EVs without isolation, brighten encapsulated RNAs, and amplify fluorescence signals in situ in a single step. We confine catalyzed hairpin DNA circuit (CHDC) in cationic lipid-polymer hybrid nanoparticles (LPHNs) that are tethered on a chip. LPHN features a core-shell-corona structure that facilitates the transfer and mixing of CHDC with EV-associated RNAs when forming the LPHN-EV nanocomplex. CHDC is triggered upon target RNA binding and quickly generate amplified signals. The signal amplification efficiency of LPHN-CHDC is demonstrated in artificial EVs, cancer cells, and cancer cell-derived EVs. We show that LPHN-CHDC biochip with signal amplification capability could selectively and sensitively identify low expression glypican-1 mRNA in serum EVs, distinguishing patients with early- and late-stage pancreatic cancer from healthy donors and patients with benign pancreatic disease.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
(3-Mercaptopropyl)trimethoxysilan, 95%
Sigma-Aldrich
3,6-Dimethyl-1,4-dioxan-2,5-dion, 99%
Sigma-Aldrich
Glykolid, ≥99%
Sigma-Aldrich
Mechlorethamin -hydrochlorid, 98%
Sigma-Aldrich
L-α-Phosphatidylethanolamin, Dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Sigma-Aldrich
1,2-Dimyristoyl-sn-glycerin, ≥95%