Skip to Content
Merck

AMBH95E088F2

Ambeed, Inc.

1-Iodo-4-isopropylbenzene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€82.05
25 G
€121.24
100 G
€255.96
500 G
€892.83

Vendor SKU: A798482-5G

€82.05


Ships in 14 days - from Aldrich Partner.


About This Item

CAS Number:
Molecular Weight:
246.09
MDL number:

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Iupac Name

1-iodo-4-isopropylbenzene

Inchi Code

1S/C9H11I/c1-7(2)8-3-5-9(10)6-4-8/h3-7H,1-2H3

InChI key

PQJOSEVTIKYWLH-UHFFFAOYSA-N

storage temp.

Keep in dark place, inert atmosphere, room temperature

Purity

95%

Country of Origin

CN

Shipping Temperature

Normal

Physical Form

liquid

Product Link

https://www.ambeed.com/products/17356-09-1.html

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

MSDS

https://www.ambeed.com/products/17356-09-1.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service