Sign In to View Organizational & Contract Pricing
Select a Size
100 MG
€206.97
250 MG
€279.23
1 G
€608.69
5 G
€1,515.00
Vendor SKU: A148240-100MG
€206.97
Ships in 14 days - from Aldrich Partner.
About This Item
Skip To
Iupac Name
4-chloropyridin-2-ylboronic acid
Inchi Code
1S/C5H5BClNO2/c7-4-1-2-8-5(3-4)6(9)10/h1-3,9-10H
InChI key
KOLVACJGJSSBEM-UHFFFAOYSA-N
storage temp.
Inert atmosphere, store in freezer, under -20C
Purity
95%
Country of Origin
CN
Shipping Temperature
Shipped in cold pack
Physical Form
solid
Product Link
https://www.ambeed.com/products/870459-91-9.html
Signal Word
Warning
Hazard Statements
Precautionary statements
MSDS
https://www.ambeed.com/products/870459-91-9.html
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service