Skip to Content
Merck

MAT047025275

Matrix Scientific

5-(Diisopropylamino)amylamine

Sign In to View Organizational & Contract Pricing

Select a Size

Product MAT047025275 is not currently sold in your country.

About This Item

CAS Number:
Molecular Weight:
186.34
MDL number:
Product MAT047025275 is not currently sold in your country.

Iupac Name

N~1~,N~1~-diisopropyl-1,5-pentanediamine

Inchi Code

1S/C11H26N2/c1-10(2)13(11(3)4)9-7-5-6-8-12/h10-11H,5-9,12H2,1-4H3

InChI key

CIFBCEXYFFKMEB-UHFFFAOYSA-N

Purity

97%

Country of Origin

US

Product Link

https://www.matrixscientific.com/007569.html

Pictograms

MSDS

https://www.matrixscientific.com/007569.html


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library