Skip to Content
Merck

BDPH9BCC4841

Bide Pharmatech

2-Bromo-6-trifluoromethylpyrazine

Synonym(s):

2-bromo-6-(trifluoromethyl)pyrazine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H2N2Br1F3
CAS Number:
Molecular Weight:
226.98
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H2BrF3N2/c6-4-2-10-1-3(11-4)5(7,8)9/h1-2H

Inchi Key

HUCJZLJSTLNVJK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under 2-Bromo-6-trifluoromethylpyrazine


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library