Skip to Content
Merck

ENA423205797

Enamine

2-[(thian-4-yl)amino]propan-1-ol

Sign In to View Organizational & Contract Pricing

Select a Size

Product ENA423205797 is not currently sold in your country.

About This Item

CAS Number:
Molecular Weight:
175.3
MDL number:
Product ENA423205797 is not currently sold in your country.

Iupac Name

2-(tetrahydro-2H-thiopyran-4-ylamino)-1-propanol

Inchi Code

1S/C8H17NOS/c1-7(6-10)9-8-2-4-11-5-3-8/h7-10H,2-6H2,1H3

InChI key

ZSTPCSRGJVKEJN-UHFFFAOYSA-N

storage temp.

RT

Purity

95

Country of Origin

UA

Shipping Temperature

Normal

Physical Form

powder

Product Link

https://www.enaminestore.com/catalog/EN300-166227

MSDS

https://www.enaminestore.com/catalog/EN300-166227


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library