description
No proteinase K or heating step needed for Magprep® Nucleic Acid Isolation,Nuclease free (NICKase, RNase, DNase), DNA-free, High-throughput Compatible and Manual
Quality Level
usage
1000 purifications
sustainability
Greener Alternative Product
concentration
45-55 mg/mL (Magnetic particle)
test parameters
total test time: 5 min
suitability
suitable for nucleic acid purification
greener alternative category
storage temp.
2-30°C
General description
The Ultra pure MagPrep® Viral RNA Lysis and Binding Buffer is intended for use within the Ultra pure MagPrep® nucleic acid purification system for manual or high throughput viral nucleic acid isolation from most biological fluids. The Ultra pure MagPrep® nucleic acid purification system is optimized for rapid easy handling and maximal yield. Nucleic acid isolation is achieved without the need for Proteinase K or sample heating, and all items are shipped and stored at ambient temperature. Buffer is quality tested to be free of contaminant nucleases and DNA, reducing the likelihood of false positives or negatives in downstream assays. Nucleic acid purified using the Ultra pure MagPrep® system is free of PCR inhibitors such as chaotropic salts or alcohols, allowing for effective amplification in various downstream applications. For research or further manufacturing use only. Not intended for direct use in humans or animals. Non-TSCA use only in US.
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of the four categories of greener alternatives. Click here to view its DfS scorecard.
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of the four categories of greener alternatives. Click here to view its DfS scorecard.
We are committed to bringing you Greener Alternative Products. This product is a Design for Sustainability (DfS) developed product, which belongs to one of the four categories of greener alternatives. Click here to view its DfS scorecard.
Features and Benefits
- Nucleic Acid Isolation
- Manual or high-throughput automated isolations
- RT-qPCR
- Digital PCR
- Biological samples can be used
- Room temperature stable reagents
- DNA free
- DNase free
- RNase free
- NICKase free
- simple and fast protocols
Legal Information
MAGPREP is a registered trademark of Merck KGaA, Darmstadt, Germany
Kit Components Only
Product No.
Description
- UPMPPART-2ML-KC Ultra pure MagPrep Magnetic Particles
- UPMPCFDBB-50ML-KC Ultra pure MagPrep cfDNA Binding Buffer
- UPMPWB1-100ML-KC Ultra pure MagPrep Wash Buffer 1
- UPMPWB2-100ML-KC Ultra pure MagPrep Wash Buffer 2
- UPMPEB-10ML-KC Ultra pure MagPrep Elution Buffer
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service