Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Quality Segment
1 of 1
This Item | |||
|---|---|---|---|
| description - | description protease substrate | description Sensitive chromogenic substrate for human leukocyte cathepsin G that is not reactive with elastase. | description - |
| Quality Level 100 | Quality Level 200 | Quality Level 100 | Quality Level - |
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



