Skip to Content
Merck

AABH97CD71EF

AA BLOCKS, INC.

17α-Hydroxypregnenolone

Synonym(s):

1-((8R,9S,10R,13S,14S,17R)-3,17-dihydroxy-10,13-dimethyl-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl)ethan-1-one

Select a Size

Change View

About This Item

Linear Formula:
C21H32O3
CAS Number:
Molecular Weight:
332.48
MDL number:
Sign in to see your pricing
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Country of Origin

US

Inchi Code

1S/C21H32O3/c1-13(22)21(24)11-8-18-16-5-4-14-12-15(23)6-9-19(14,2)17(16)7-10-20(18,21)3/h4,15-18,23-24H,5-12H2,1-3H3/t15?,16-,17+,18+,19+,20+,21+/m1/s1

Inchi Key

JERGUCIJOXJXHF-QUPIPBJSSA-N

Storage temp.

2~8C





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library