Skip to Content
Merck

AABH97D205BD

AA BLOCKS, INC.

Fast Red Violet LB Salt

Synonym(s):

4-(benzoylamino)-5-chloro-2-methylbenzenediazonium chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C14H11O1N3Cl2
CAS Number:
Molecular Weight:
308.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C14H10ClN3O.ClH/c1-9-7-13(11(15)8-12(9)18-16)17-14(19)10-5-3-2-4-6-10;/h2-8H,1H3;1H

Inchi Key

SARKXLKWFKNUMR-UHFFFAOYSA-N

Purity

90%

Storage temp.

-20C

Country of Origin

US





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library