Skip to Content
Merck

GF25289837

Zinc

foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%

Synonym(s):

Zinc, ZN000235

Sign In to View Organizational & Contract Pricing

Select a Size

Pricing and availability is not currently available.

About This Item

Empirical Formula (Hill Notation):
Zn
CAS Number:
Molecular Weight:
65.39
MDL number:
UNSPSC Code:
12141750
PubChem Substance ID:
NACRES:
NA.23

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

vapor pressure

1 mmHg ( 487 °C)

Assay

99.99%

form

foil

manufacturer/tradename

Goodfellow 252-898-37

resistivity

5.8 μΩ-cm, 20°C

diam. × thickness

8 mm × 0.05 mm

bp

907 °C (lit.)

mp

420 °C (lit.)

density

7.133 g/mL at 25 °C (lit.)

SMILES string

[Zn]

InChI

1S/Zn

InChI key

HCHKCACWOHOZIP-UHFFFAOYSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF19068037GF44141712GF61733101
Zinc foil, 8mm disks, thickness 0.05mm, as rolled, 99.99+%

GF25289837

Zinc

Zinc foil, 8mm disks, thickness 0.25mm, as rolled, 99.99+%

GF19068037

Zinc

Zinc foil, 8mm disks, thickness 0.5mm, as rolled, 99.99+%

GF44141712

Zinc

Zinc foil, 8mm disks, thickness 0.05mm, as rolled, 99.95+%

GF61733101

Zinc

assay

99.99%

assay

99.99%

assay

99.99%

assay

99.95%

form

foil

form

foil

form

foil

form

foil

manufacturer/tradename

Goodfellow 252-898-37

manufacturer/tradename

Goodfellow 190-680-37

manufacturer/tradename

Goodfellow 441-417-12

manufacturer/tradename

Goodfellow 617-331-01

mp

420 °C (lit.)

mp

420 °C (lit.)

mp

420 °C (lit.)

mp

420 °C (lit.)

resistivity

5.8 μΩ-cm, 20°C

resistivity

5.8 μΩ-cm, 20°C

resistivity

5.8 μΩ-cm, 20°C

resistivity

5.8 μΩ-cm, 20°C

bp

907 °C (lit.)

bp

907 °C (lit.)

bp

907 °C (lit.)

bp

907 °C (lit.)

General description

For updated SDS information please visit www.goodfellow.com.

Legal Information

Product of Goodfellow

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Huibo Chen et al.
Nanoscale research letters, 5(3), 570-575 (2009-01-01)
We reported the optical and wettability properties of aligned zinc oxide micro/nanotube arrays, which were synthesized on zinc foil via a simple hydrothermal method. As-synthesized ZnO micro/nanotubes have uniform growth directions along the [0001] orientations with diameters in the range
Hongqin Liu et al.
ACS applied materials & interfaces, 1(9), 2086-2091 (2010-04-02)
Zinc, silicon, and steel superhydrophobic surfaces were prepared by a simple solution-immersion technique. In the case of zinc, the method consists of dipping of the substrate in a prehydrolyzed methanol solution of 1H,1H,2H,2H-(perfluorooctyl)trichlorosilane [CF(3)(CF(2))(5)(CH(2))(2)SiCl(3), PFTS] for 24 h at 50
Pritesh Hiralal et al.
ACS nano, 4(5), 2730-2734 (2010-04-27)
Solid-state and flexible zinc carbon (or Leclanche) batteries are fabricated using a combination of functional nanostructured materials for optimum performance. Flexible carbon nanofiber mats obtained by electrospinning are used as a current collector and cathode support for the batteries. The
Chenglin Yan et al.
The journal of physical chemistry. B, 110(51), 25850-25855 (2006-12-22)
Vertically aligned ZnO nanorods with uniform diameter and length have been synthesized on a zinc foil substrate with ammonium persulfate as oxidant via a facile, larger scale production and inexpensively synthesized method without any templates or additives. SEM and XRD
Chuanhui Xia et al.
Journal of nanoscience and nanotechnology, 11(12), 10506-10510 (2012-03-14)
We have investigated the properties of Mn-doped ZnO nanocrystalline film growing on zinc foil by the hydrothermal method. X-ray photoelectron spectroscopy shows that the manganese ions exist as Mn2+ in the film. From UV-vis spectra, we observe a red shift

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service