Skip to Content
Merck

Skip To

ATE428009727

AstaTech, Inc.

PIMECROLIMUS

Synonym(s):

(3S,4R,5S,8S,12R,14S,15R,16S,18R,19R,E)-3-((E)-1-((1R,3R,4S)-4-chloro-3-methoxycyclohexyl)prop-1-en-2-yl)-8-ethyl-5,19-dihydroxy-14,16-dimethoxy-4,10,12,18-tetramethyl-5,6,8,11,12,13,14,15,16,17,18,19,24,25,26,26a-hexadecahydro-3H-15,19-epoxypyrido[2,1-c][1]oxa[4]azacyclotricosine-1,7,20,21(4H,23H)-tetraone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg
Ships in 14 days - from Aldrich Partner.
€92.63
100 mg
Ships in 14 days - from Aldrich Partner.
€121.39
250 mg
Ships in 14 days - from Aldrich Partner.
€285.53

Vendor SKU: 41186-25MG

€92.63


Ships in 14 days - from Aldrich Partner.

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Country of Origin

CN

Inchi Code

1S/C43H68ClNO11/c1-10-30-18-24(2)17-25(3)19-36(53-8)39-37(54-9)21-27(5)43(51,56-39)40(48)41(49)45-16-12-11-13-32(45)42(50)55-38(28(6)33(46)23-34(30)47)26(4)20-29-14-15-31(44)35(22-29)52-7/h18,20,25,27-33,35-39,46,51H,10-17,19,21-23H2,1-9H3/b24-18+,26-20+/t25-,27-,28-,29+,30+,31+,32?,33+,35-,36+,37+,38-,39-,43-/m1/s1

Inchi Key

KASDHRXLYQOAKZ-XRYIWSAPSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

2-8 C

Physical Form

White to Yellow Solid


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
1539100SML1437
description

-

description

United States Pharmacopeia (USP) Reference Standard

description

≥97% (HPLC)

Physical Form

White to Yellow Solid

Physical Form

-

Physical Form

-

Inchi Key

KASDHRXLYQOAKZ-XRYIWSAPSA-N

Inchi Key

-

Inchi Key

-

Purity

95%

Purity

-

Purity

-

Shipping Temperature

Normal

Shipping Temperature

-

Shipping Temperature

-

Storage temp.

2-8 C

Storage temp.

-

Storage temp.

-

Country of Origin

CN

Country of Origin

-

Country of Origin

-


Still not finding the right product?

Explore all of our products under PIMECROLIMUS


hstatements

Precautionary Statements

Signal Word

Warning



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library