Skip to Content
Merck

BDPH9BCC28F3

Bide Pharmatech

2-Bromothiophene-3-carbaldehyde

Synonym(s):

2-bromo-3-thiophenecarbaldehyde

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H3O1Br1S1
Molecular Weight:
191.05
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H3BrOS/c6-5-4(3-7)1-2-8-5/h1-3H

Inchi Key

MMEGVQIGIBCTHI-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library