Skip to Content
Merck

BDPH9BCC2ACA

Bide Pharmatech

2-Amino-6-methylpyrimidine-4-thiol

Synonym(s):

2-amino-6-methyl-4(3H)-pyrimidinethione

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H7N3S1
CAS Number:
Molecular Weight:
141.2
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H7N3S/c1-3-2-4(9)8-5(6)7-3/h2H,1H3,(H3,6,7,8,9)

Inchi Key

VWMGCVRJTGJRSS-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?

Explore all of our products under 2-Amino-6-methylpyrimidine-4-thiol


Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library