Skip to Content
Merck

BDPH9BCC41F3

Bide Pharmatech

Ethyl 1-(aminomethyl)cyclopropanecarboxylate

Synonym(s):

ethyl 1-(aminomethyl)cyclopropanecarboxylate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H13O2N1
CAS Number:
Molecular Weight:
143.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H13NO2/c1-2-10-6(9)7(5-8)3-4-7/h2-5,8H2,1H3

Inchi Key

AYTOKAZVIPSHHI-UHFFFAOYSA-N

Shipping Temperature

Shipped in cold pack

Purity

98%

Storage temp.

Keep in dark place, sealed in dry, store in freezer, under -20C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Danger

Pictograms

FlameCorrosion


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library