Skip to Content
Merck

BDPH9BCC52BE

Bide Pharmatech

3,3'-Difluoro-[1,1'-biphenyl]-4,4'-dicarboxylic acid

Synonym(s):

3,3'-difluoro[1,1'-biphenyl]-4,4'-dicarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C14H8O4F2
CAS Number:
Molecular Weight:
278.21
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C14H8F2O4/c15-11-5-7(1-3-9(11)13(17)18)8-2-4-10(14(19)20)12(16)6-8/h1-6H,(H,17,18)(H,19,20)

Inchi Key

PSAXGLIYDQKPDK-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library