Skip to Content

BDPH9BCC61EA

Bide Pharmatech

5-(Trifluoromethyl)quinolin-8-amine

Synonym(s):

5-(trifluoromethyl)-8-quinolinamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C10H7N2F3
CAS Number:
Molecular Weight:
212.17
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C10H7F3N2/c11-10(12,13)7-3-4-8(14)9-6(7)2-1-5-15-9/h1-5H,14H2

Inchi Key

KJLQGKLEIKGMCQ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Solid



Still not finding the right product?


hstatements

Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library