Skip to Content
Merck

Skip To

BDPH9BCC673D

Bide Pharmatech

6-Chloro-1-methylpyrazin-2(1H)-one

Synonym(s):

6-chloro-1-methylpyrazin-2(1H)-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H5O1N2Cl1
Molecular Weight:
144.56
MDL number:
Product BDPH9BCC673D is not currently sold in your country.


Inchi Code

1S/C5H5ClN2O/c1-8-4(6)2-7-3-5(8)9/h2-3H,1H3

Inchi Key

PYOXFHZKMCUREC-UHFFFAOYSA-N





Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library