Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available. Contact Technical Service
Inchi Code
1S/C10H6O/c1-2-8-3-4-10-9(7-8)5-6-11-10/h1,3-7H
Inchi Key
NFOIHYYIUMHKKC-UHFFFAOYSA-N
Shipping Temperature
Cold-chain transportation
Purity
95%
Storage temp.
Sealed in dry, 2-8C
Country of Origin
CN
Physical Form
Liquid
Still not finding the right product?
Explore all of our products under 5-Ethynylbenzofuran
hstatements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
