Skip to Content
Merck

BDPH9BCC7A2F

Bide Pharmatech

7-Amino-3,4-dihydroisoquinolin-1(2H)-one

Synonym(s):

7-amino-3,4-dihydro-1(2H)-isoquinolinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C9H10O1N2
CAS Number:
Molecular Weight:
162.19
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C9H10N2O/c10-7-2-1-6-3-4-11-9(12)8(6)5-7/h1-2,5H,3-4,10H2,(H,11,12)

Inchi Key

VNUQVKZHEBCTNT-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library