Skip to Content
Merck

BDPH9BCC8A62

Bide Pharmatech

2-Chloro-4-methyl-6-(trifluoromethyl)pyridine

Synonym(s):

2-chloro-4-methyl-6-(trifluoromethyl)pyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C7H5N1Cl1F3
CAS Number:
Molecular Weight:
195.57
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C7H5ClF3N/c1-4-2-5(7(9,10)11)12-6(8)3-4/h2-3H,1H3

Inchi Key

KAYVWTDOIYYSDL-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid or Solid or Semi-solid



Still not finding the right product?


Precautionary Statements

Signal Word

Warning

Pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library