Skip to Content
Merck

BDPH9BCCBF8A

Bide Pharmatech

4-(Cyclopropylmethoxy)-3-fluorobenzoic acid

Synonym(s):

4-(cyclopropylmethoxy)-3-fluorobenzoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C11H11O3F1
CAS Number:
Molecular Weight:
210.2
MDL number:
Product BDPH9BCCBF8A is not currently sold in your country.


Inchi Code

1S/C11H11FO3/c12-9-5-8(11(13)14)3-4-10(9)15-6-7-1-2-7/h3-5,7H,1-2,6H2,(H,13,14)

Inchi Key

QSUMQCBKOZDHNX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

White to yellow powder or crystals



Still not finding the right product?



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library