Shipping Temperature
Normal
Purity
97%
Storage temp.
Keep in dark place, sealed in dry, room temperature
Country of Origin
CN
Physical Form
Form: Crystal - Powder
Inchi Code
1S/C6H7F3N4.ClH/c7-6(8,9)5-12-11-4-3-10-1-2-13(4)5;/h10H,1-3H2;1H
Inchi Key
AQCSCRYRCRORET-UHFFFAOYSA-N
Still not finding the right product?
Explore all of our products under 3-(Trifluoromethyl)-5,6,7,8-tetrahydro-[1,2,4]triazolo[4,3-a]pyrazine hydrochloride
hstatements
Precautionary Statements
Signal Word
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
