Skip to Content
Merck

BDPH9BCCDFB3

Bide Pharmatech

Ethyl 3,3,3-trifluoropropanoate

Synonym(s):

ethyl 3,3,3-trifluoropropanoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C5H7O2F3
CAS Number:
Molecular Weight:
156.1
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Inchi Code

1S/C5H7F3O2/c1-2-10-4(9)3-5(6,7)8/h2-3H2,1H3

Inchi Key

FMDMKDPUFQNVSH-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

98%

Storage temp.

Sealed in dry, 2-8C

Country of Origin

CN

Physical Form

Colorless to light-yellow liquid



Still not finding the right product?

Explore all of our products under Ethyl 3,3,3-trifluoropropanoate


Precautionary Statements

Signal Word

Danger

Pictograms

FlameExclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library